BR73203-EN - Ion Chromatography Troubleshooting Guide
The Ion Chromatography Troubleshooting Guide addresses common IC issues such as sample injection problems, retention time variations, detection anomalies, peak shape issues, selectivity challenges, and suppression module errors. Solutions include checking eluent and sample preparation, ensuring proper equipment maintenance, and verifying system settings. Regular servicing and adherence to manufacturer guidelines are crucial for optimal performance.
Description:
Ion Chromatography (IC) is the premier technique for determining ionic compounds in solution. The basic components consist of an eluent source, pump, sample injector, separating column, suppressor, and detector. As with any lab instrumentation, the IC system should be serviced regularly to ensure peak performance. If problems arise that are not addressed by routine maintenance, the attached troubleshooting guide provides suggested solutions for quickly resuming operation.
Environment:
Thermo Scientific™ Dionex™ IC systems
Attachment(s):
| File | Last Modified |
|---|---|
| BR73203-EN - Ion Chromatography Troubleshooting Guide.pdf | June 09, 2022 |