Thermo Scientific Dionex ICS-6000 HPIC System - Production Specifications
The Thermo Scientific Dionex ICS-6000 HPIC System is a high-pressure ion chromatography system designed for increased productivity and performance. It supports capillary, microbore, and standard bore formats, operating up to 5000 psi, and features modular design, reagent-free technology, and advanced software for ease of use and adaptability. The system includes consumables tracking, remote monitoring, and various detectors and pumps, making it versatile for a wide range of applications.
Description:
The Thermo Scientific Dionex ICS-6000 HPIC System is a high-pressure ion chromatography system that provides an impressive combination of increased productivity, expanded capabilities, and improved performance. Modular versatility, functional integration, superior performance and the capability to operate continuously at up to 5000 psi culminate in the most advanced ion chromatography system.
Environment:
- Thermo Scientific Dionex ICS-6000 HPIC System
Attachment(s):
| File | Last Modified |
|---|---|
| Thermo Scientific Dionex ICS-6000 HPIC System - Production Specifications.pdf | January 05, 2022 |