065215 - Rev 04 - Dionex ICS-900 Ion Chromatography System Operators Manual
Description:
The Thermo Scientific Dionex ICS-900 Ion Chromatography System Dionex ICS-900 performs isocratic ion analyses using suppressed conductivity detection. The Dionex ICS-900 is an integrated ion chromatography system consisting of a pump, an injection valve, and a conductivity cell.
Environment:
- Thermo Scientific Dionex ICS-900 Ion Chromatography System Dionex
Attachment(s):
File | Last Modified |
---|---|
065215 - Rev 04 - Dionex ICS-900 Ion Chromatography System Operators Manual.PDF | December 15, 2021 |