Thermo Fisher Scientific
032319 - Rev 03 - High-Pressure Inline Filter
The Dionex High-Pressure Inline Filter Installation Instructions detail the setup and maintenance of the inline filter (P/N 044105) for eluent and sample filtration. The filter removes particulates down to 0.45 microns. Proper installation involves connecting the filter to the pump and injection valve, checking for discoloration, and replacing filter disks when needed. The document emphasizes careful handling of parts to ensure a proper seal.
Description: 
The Dionex High-Pressure Inline Filter (P/N 044105) provides pre-column filtering to remove particulates down to 0.45 micron from samples or eluent.
Environment:
Dionex High-Pressure Inline Filter (P/N 044105)
  • Was this article helpful?