Thermo Fisher Scientific
065069-04 - Carbonate Removal Device 200 CRD-200 Columns - Quick Start
The document is a Quick Start guide for the Carbonate Removal Device 200 (CRD-200) Columns, emphasizing careful handling and proper operation. It includes steps for hydrating the CRD-200 using syringes and Luer adaptors to push degassed deionized water through specified ports, followed by a 10-minute hydration period. It warns against disassembling the device and exceeding 100 psi total backpressure, which could cause irreversible damage.
Description: 
This quick start guide contains the information concerning:
  • Hydrating the CRD-200
  • Backpressure Instructions
Environment:
  • Carbonate Removal Device 200
  • Was this article helpful?