065213 - Rev 01 - Carbonate Removal Device CRD 300 Product Manual
The CRD 300 Product Manual details the design, installation, and operation of the Carbonate Removal Device (CRD 300) for use in ion chromatography. The CRD 300 removes CO2 from the eluent stream to reduce background conductivity and improve analyte sensitivity.
Description:
The Carbonate Removal Device 200 and 300 CRD 200 and CRD 300 are membrane-based modules that transport CO2 from the eluent stream into the outer regenerant waste stream for removal.
Environment:
- CRD 300 (4 mm) (P/N 064637)
- CRD 300 (2 mm) (P/N 064638)
Attachment(s):
| File | Last Modified |
|---|---|
| 065213 - Rev 01 - Carbonate Removal Device CRD 300 Product Manual.PDF | January 05, 2022 |