065245-01 - Carbonate Removal Device 300 CRD-300 - Quick Start
The attachment provides quick start instructions for the Carbonate Removal Device (CRD-300). The guide provides steps for hydrating the CRD-300 and setting the appropriate backpressure. Backpressures in excess of 100 psi must be avoided.
Description:
This quick start guide contains the information concerning:
- Hydrating the CRD-300
- Backpressure Instructions
Environment:
- Carbonate Removal Device 300
Attachment(s):
| File | Last Modified |
|---|---|
| 065245-01 - Carbonate Removal Device 300 CRD-300 - Quick Start.PDF | December 30, 2021 |