Thermo Fisher Scientific
50133369 - Thermo Scientific Dionex IC Pure Water Purification System - Operating Instruction Manual
The Thermo Scientific Dionex IC Pure Water Purification System manual details its operation, including installation, safety precautions, and maintenance. The system purifies water for laboratory use, producing ASTM Type I and II water. It features modes for continuous operation, interval operation, and integration with IC systems. The manual provides instructions for connecting the system to various Dionex IC models, setting up external tanks, and performing maintenance tasks.
Description: 
Operating instruction manual
Environment:
Thermo Scientific Dionex IC Pure Water Purification System, models:
  • Dionex IC Pure System with pretreatment cartridge
  • Dionex IC Pure System without pretreatment cartridge