PSB-364 - IC OQ-PQ Rev 4.00
There is a bug in the Rev 4.00 release of the IC OQ/PQ templates for amperometry detectors using tri-mode drivers. This bug causes report calculations to remain blank, displaying "N/A." The issue affects only tri-mode drivers with amperometry detection. The bug will be fixed in IC OQ/PQ Rev 4.01, included on the PeakNet 6.40 SP1 CD. The kits will not be updated with the new version, which will be available solely on the 6.40 SP1 CD.
Description:
There is a bug in the Rev 4.00 release of the IC OQ-PQ templates for amperometry DC or Integrated detectors when using the new tri-mode drivers. IC OQ-PQ Rev 4.01 will be released and included on the PeakNet 6.40 SP1 CD to fix this bug. The kits will not be updated with the new version. It will only be available on the 6.40 SP1 CD.
Environment:
- IC OQ-PQ Rev 4.00 templates for amperometry DC or Integrated detectors
- IC OQ-PQ Rev 4.01
- PeakNet 6.40 SP1 CD
- All SN
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-364 - IC OQ-PQ Rev 4.00.PDF | January 07, 2022 |