PSB-399 - IC OQ-PQ Rev 4.04a
IC OQ/PQ Rev 4.04a was released on June 27, 2003, and is included in all relevant validation kits and the June 2003 Chromeleon/PeakNet Service Pack. This update fixes issues with the AS50 running CM 6.5 with Sample Prep Overlap enabled, corrects the OQ/PQ IC report, and updates various template commands.
Description:
IC OQ-PQ Rev 4.04a was released effective June 27 2003. All shipments of the IC OQ-PQ validation kits PN 057599 Kit with Test cells and PN 057608 Basic Refill Kit will contain Rev 4.04a. The June 2003 Chromeleon-PeakNet Service Pack also contains IC OQ-PQ 4.04a.
Environment:
- IC OQ-PQ Rev 4.04a
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-399 - IC OQ-PQ Rev 4.04a | January 07, 2022 |