Thermo Fisher Scientific
AS-AP Error: Alignment positions not precise
Error Code
"Alignment positions not precise" and other errors related to the XZ drive on the AS-AP
Environment
 AS-AP
Cause
During the needle alignment procedure, an error occurs  when performing the regular self-test. The self test fails when the needle goes back over to the inject port, and before it goes over to the wash port. (Refer to the video in the attachments)
Resolution Steps
1. Reset the AS-AP to default alignment parameters: 
https://knowledge1.thermofisher.com/Chromatography/Ion_Chromatography/Ion_Chromatography_Knowledge/Resetting_the_AS-AP_alignment_parameters
Resetting_the_AS-AP_alignment_parameters.pdf
2. Set the alignment parameters to some default/example values (snip from PSB-ICS_-1079_ASAP-XZ Drive Issues, attached)
clipboard_e052462edb0e5eb97db8db20d44385541.png
PSB-ICSP-1079_ASAP-XZ Drive Issues.pdf
    3. Additional set of default coordinates:
clipboard_e26385e8d1f06a52336e7d03633270de8.png
  • Was this article helpful?