Dionex™ PC10 Postcolumn Pneumatic Delivery System
Issue
Cannot locate manual or documents related to PC10.
Reduced flow rate.
Regulator leaks.
Environment
PC10 pneumatic reagent delivery system used with ICS-6000, ICS-5000+ or similar
Resolution
Files below include:
- Automation Kit Installation Instructions
- PC10 Assy Drawing
- Pressure Vessel Assy drawing
- Reagent Organizer drawing
- Reagent Organizer Rack drawing
- Transition Metal System with PC10 Installation and Troubleshooting Manual
- Public website landing page
To address reduced flow rate, recommended to use a 250 mL bottle rather than the 1 L bottle.
If the regulator leaks, then order P/N 035971 (RGLTR,0-100PSI) through your local Order Processing Team. It looks like this:
Attachment(s)
| File | Last Modified |
|---|---|
| PC10 post column delivery system install.pdf | December 22, 2022 |
| PC10.docx | September 02, 2022 |
| PC10 - Reagent Organizer Rack - 050602.pdf | September 02, 2022 |
| PC10 - Reagent Organizer - 050802.pdf | September 02, 2022 |
| PC10 - Pressure Vessel Assy - 037460.pdf | September 02, 2022 |
| PC10 - Post-Column Pneumatic Controller - 043903.pdf | September 02, 2022 |
| PC10 - Automation Kit Installation Instructions.pdf | September 02, 2022 |
| PC10 regulator.png | September 02, 2022 |