Extending the Inuvion Eluent monitor USB cable
Issue
When a brand new Inuvion Eluent Monitor is shipped it comes with the USB cable coiled inside the base plate. This USB cable can be extended in length by simply removing the base plate and extending the coiled USB cable.
Environment
P/N 22185-62707 Inuvion Eluent monitor for 2-litre eluent bottle.
P/N 22185-62708 Inuvion Eluent monitor for 4-litre eluent bottle.
Resolution
The attached Knowledge Article has step by step instructions on how to extend the Inuvion Eluent Monitor USB cable by removing the base plate.
Attachment(s)
| File | Last Modified |
|---|---|
| Extending the Inuvion Eluent monitor USB cable.pdf | July 10, 2024 |