How Do I troubleshoot a Leaking Suppressor?
Issue:
IC suppressor is leaking due to possible external back pressure.
Environment:
IC Suppressor
Resolution:
- Check the installation guide for the proper configuration.
- Install the correct back pressure coils and check the back pressure.
Cause:
Incorrect back pressure coils were used.