ICS Conductivity Detector Letter of Equivalence
Issue
How come part number 061830 (ASSY,CD DET/CELL,ANALYTICAL,ICS3/5) has been superseded with part number 079829 (CD Conductivity Detector, analyt., DC).?
Why can't I order part number 061830 (ASSY,CD DET/CELL,ANALYTICAL,ICS3/5)?
I need a Letter of Equivalence (LOE) for my records to state newer part has same fit, form and function.
Resolution
See attached generic customer Letter of Equivalence.
If a Customized letter is required, then please request from the IC/SP PSE Team, icsp.productsupport@thermofisher.com.
- The Customer name, company name, address, and phone number are required to provide a customized LOE.
Attachment(s)
| File | Last Modified |
|---|---|
| CD_LOE.pdf | August 09, 2022 |