Integrion experiencing disconnection issues
Issue
IC products, mostly Integrion have been experiencing disconnection issues.
Environment
Integrion
Resolution
After receiving a computer, verify the following:
- All the cables are connected.
- Windows has been activated.
- Chromeleon has been installed including IQ and OQ.
- Instrument is connected to computer.