Thermo Fisher Scientific
Inuvion Pressure Transducer Parts and Fittings
Issue
Fittings/parts on the Inuvion pump/pressure tranducer unit that can be ordered separately from the complete assembly.
Environment
 Inuvion
Resolution
The parts listed below can be ordered separately from the complete pressure transducer assembly. If other parts in this assembly are needed, please order the complete assembly (part B51002664). Pressure Transducer Assembly (part B51002664) drawing from Inuvion Service Manual):
clipboard_e23c381205923f83a9d49c9c66e17e43e.png
The seal for the purge valve screw is the part labeled 8 in snip above: 063382, SEAL,WASH,DURON 14 GOLD,50809,RoHS 
Barb fitting  labeled 5 in the snip above is part 035528. 
Purge valve screw (knob) is part 3 in the snip above: 22185-20204