RFID consumable tracking error
Issue
Sequence cannot start or running sequence is interrupted due to consumable scan issue in Integrion, error code #230, #235 and #240, “RFID label read/write error."
Environment
- Integrion
- Chromeleon
- RFID
Resolution
- Access to the Consumables Inventory Page in Chromeleon software, check if all the RFID devices are identified. If yes, execute Rescan and Approve, if not, go to step 2
- Check all the connections of one-wire devices and columns, make sure all the one-wire connectors are securely connected and tightly screwed.
- Open the door of the column oven, check the RFID tags: the labels for separation and guard columns must not be wrapped around a column or pressed flush against an internal surface.
- Make sure RFID labels are not caught in the compartment door and do not touch bare metal, including the heat exchanger.
- Confirm that RFID labels on the separation and guard columns must not be parallel or stacking on top of each other.
- Execute a Rescan and Approve the consumables, make sure After clicking Approve, do not close the window. Wait for the Rescan button to become active & the Approve button to remain inactive. This process could take up to one minute.
- If the sequence ready check still fails after rescanning and re-approving the consumables, or the sequence running could be started but will be interrupted during the running caused by RFID Read/Write failures, then replace the RFID Interface board (P/N 22153-98003), for temporary use, go to step 9 to disable the RFID label scan function manually.
- If step 7 does not resolve the issue, replace the three RFID Antenna A, B, C (P/N 22153-98004, P/N 22153-98005, P/N 22153-98006).
- Manually disable the RFID label scan function: No software button in Chromeleon to disable the RFID label scanning function, open the left panel of Integrion and locate the RFID interface board at the upper right position and its 8-pin white connector in the middle right of the main board. Disconnect this RFID interface board from the mainboard. No RFID tags could be detected and must be scanned and approved.
- Reconnect all the one wire RFID devices and RFID label devices. Make sure RFID labels are not caught by the door, not pressed, not touch anything, and not be parallel and stacking. Wait longer after clicking the Re-scan then Approve button. If all the actions don’t work, replace the RFID Interface board and RFID Antenna A, B, C. For urgent and temporary use, disconnect the RFID Interface board cable from the mainboard to disable the RFID label scan function.