Stop slow carousel rotation on AS-AP autosampler
Issue
While measuring the carousel tray temperature during a qualification, it may be necessary to temporarily stop the carousel/tray rotation.
Environment
AS-AP with temperature control
Resolution
1. Open Chromeleon Console, Instrument Category
2. Go into F8/Commands. The command is visible on the Commands Tab under any level.
3. Under the Sampler Commands:
It is also possible to enter the command in an instrument method script editor:
The RotationMode=Start command can be entered after the testing.