Thermo Fisher Scientific
Suppressor Not Recognized In Chromeleon
Issue
Suppressor containing one-wire chips not being recognized in Chromeleon for Integrion and ICS-6000. 
Environment
  • IC Suppressors
  • ICS-6000
  • Integrion
Resolution
Please contact your local customer care.
Analysis
There are two possible errors when plugging in a suppressor that was not programmed into an Integrion or ICS-6000.
The first error observed is when the suppressor is connected to Chromeleon, the user will receive a “Code 25: Wrong device connected to connector A” error in the audit trail, as seen in Figure 1.
Error Code 25.png
Figure 1. Code 25: Wrong device connected to connector A error.
The second error observed is that the user can receive is when the suppressor is connected to Chromeleon, it will show up as a “ERD_2mm” device with the serial number “123456789” as see in Figure 2.
ERD_2mm device.png
Figure 2. ERD_2mm device with serial number 123456789.
Cause
The errors are caused by the suppressor not being programmed correctly.