Syringe Drive Error
Issue
Syringe drive error due to a loose nut which holds the lead screw. There is a missing a blue loctite to keep nut secure.
Environment
AS-AP Autosampler
Resolution
- Perform a quality check on instruments and parts often to prevent future malfunctions due to poor quality, defective and/or missing parts.
- Report any findings to have repaired or replaced.