USB disconnect on Integrion
Issue
- USB disconnect of Integrion
- USB disconnect of modules
- Loss of communication
- Timeout read response error
- U0000 CD Data
Environment
- Integrion
- ICS-6000 DC
- ICS-5000 DC
- ICS-5000+ DC
- Dell Computers
- HP Computers
- Chromeleon 7.x
- Windows 7 or Windows 10
Resolution
Below is a list of recommended troubleshooting steps (superseding steps outlined in PSB-1105). This list is for guidance only and one may have to perform multiple measures to resolve the issue. Finally, not all steps work the same for each case, or report.
- Optimize computer Bios, update drivers and update Windows setting according to https://knowledge1.thermofisher.com/@go/path/disconnect.
- Optimize power settings per Factory Communication # 2012.044. Oftentimes, PC manufacturers will default to a power plan that is not “always on”
- Implement PSB 1084.
- Escalate to IC/SP PSE Team (icsp.productsupport@thermofisher.com).
- If applicable and possible, remove the PC from the Customer’s network. There have been a few instances where the Customer’s IT policies and updates were reverting the PC power settings to the default settings.
- Order the StarTech 4 port PCI USB card and StarTech Mini 4-pin Floppy FDD Female (SP4) to 15-pin SATA Male Power Adapter Cable.
Example images of installation of StarTech 4 port PCI USB card and StarTech Mini 4-pin Floppy FDD Female (SP4) to 15-pin SATA Male Power Adapter Cable
- Order the StarTech 4-port externally powered USB 2.0 hub, P/N 060392 (HUB,USB,4PORT,STARTECH,RoHS).
- Replace the PC.
Cause
Root cause analysis is ongoing by ICSP.
Attachment(s)
| File | Last Modified |
|---|---|
| FC_2020.006_-_Rev_B_-_Factory_Computer_Settings_for_W10-LTSB_and_W10-LTSC (002).pdf | September 18, 2023 |
| USB Cable.png | February 24, 2022 |
| StarTech.png | February 23, 2022 |