USB Disconnection Issues
Issue
How do I troubleshoot disconnection issues with my Dell computer?
Environment
- IC systems
- IC modules
- Dell and non-Dell computers
Resolution
| What to Check | How to Check |
|
1. To bring up the Run dialog box, press Windows key + R key. 2. In the run dialog box, type regedit to access the Registry Editor. 3. Click on: HKEY_LOCAL_MACHINE\HARDWARE\DESCRIPTION\BIOS to see the BIOS settings and version. |
|
1. Navigate to Dell Support Homepage 2. Input Service Tag number. This number will be located on your computer. 3. Select SEARCH. 4. Select 'Drivers & Downloads' located in the center of your screen below your computer model details. 5. Locate the "Manually find a specific driver for your computer" and click on FIND DRIVERS. 6. Select "Show downloads for only THIS PC..." NOTE: Do not use the "Let Dell automatically find driver updates for you" option. 6. A comprehensive list of all available uploads will appear. 7. Select the ACTION you wish to proceed with by clicking on DOWNLOAD. 8. Recommendations on how to update/download:
|
|
1. Click the Start or Windows button, located usually in the lower-left corner of your computer screen. 2. Click Settings. 3. Click System. 4. Click About located in the lower left of the screen. The resulting screen shows the edition of Windows. 5. Confirm Windows version under 'Windows Specifications.' 6. Click Check for updates and update as needed. |
|