Using the Rear Panel Relay on an Inuvion
Issue
Relay on an Inuvion is not sending the start signal for the A2D box.
Environment
Inuvion
Resolution
Uncheck the Zero Flow Indicator Box in the Inuvion instrument configuration. Once this box is UNCHECKED, then the relay will perform other functions (i.e. start signal for an external detector or A2D box).
Using the green terminal block on the back of the Inuvion, connect the black wire to "Relay-COM" and the red wire to "Relay-NO" to send a start signal.
The Relay command to close the relay will need to be added to the instrument method.