VWD-IC Critical Module Failures
Issue
IC VWD Critical Module Failures, as well as, ADC internal test failure
Environment
Ion Chromatography
Resolution
Step 1: Assess what errors are showing in Chromeleon audit trail. If “critical module failure”, then go to step 2. If “ADC internal test failure”, then go to step 4.
Step 2: If “critical module failure” error, then replace mainboard, transfer old jumpers from old mainboard to new mainboard, and run F Test per ICS Series VWD Service Manual.
Step 3: If “critical module failure” error persists, then replace optics bench (See PSB-IC-SYS-1025).
Step 4: If “ADC internal test failure” error appears, then try the following actions in this order. 1) Turn off module 2) Remove Power cable 3) Remove USB cable 4) Wait 5 minutes 5) Reconnect power and USB cables and turn on.
Step 5: If “ADC internal test failure” error persists, then replace optics bench (See PSB-IC-SYS-1025).
Step 6: If none of the above are resolved or further issues are encountered, then please contact IC/SP PSE (icsp.productsupport@thermofisher.com) for options such as an RMA repair or warranty replacement/exchange.
Analysis
Parts required:
| Quantity | Part Number | Description |
| 1 | 6074.3001 | Mainboard (Main board, completely assembled (VWD Single Channel) |
| 1 | 6074.3010 | Main board, completely assembled (VWD Multiple Channel) |
| 1 | 6074.3012 | Replacement Mirror Kit |
| 1 | 6309.0263 | Optics Cleaning Tissues |
| 1 | 6074.1000 | Replacement Optical Unit, Single Wavelength VWD |
| 1 | 6074.1010 | Replacement Optical Unit, 4 Channel VWD) |
Attachment(s)
| File | Last Modified |
|---|---|
| VWD-IC critical module failures - image.jpg | January 11, 2022 |