D07669 - Rev - 1.1 - Qualification of Thermo Scientific Multidrop 384 and DW Unit
The Thermo Scientific Multidrop® 384/DW unit requires qualification protocols for installation, operation, and performance. Installation involves verifying conditions, setup, and user training. Operational qualification tests functionalities and calibration. Performance tests accuracy and precision using specific equipment and reagents. Customers provide records, workspace, and approve documents promptly. Warranty covers repair and re-qualification costs under certain conditions.
Description:
Qualification and specification sheet
Environment:
- Thermo Scientific Multidrop® 384
- Thermo Scientific Multidrop® DW
Attachment(s):
| File | Last Modified |
|---|---|
| D07669 - Rev - 1.1 - Qualification of Thermo Scientific Multidrop 384 and DW Unit.pdf | January 07, 2022 |