NO5616 - Rev 1.6 - Thermo Scientific Multidrop Combi - User Manual
The Thermo Scientific Multidrop Combi User Manual provides instructions for the use, maintenance, and troubleshooting of the Multidrop Combi microplate dispenser. It emphasizes safety precautions, proper setup, and routine maintenance to ensure optimal performance. The instrument supports dispensing into 96, 384, and 1536-well plates, offering flexibility with a volume range of 0.5 to 2500 µl. The manual also highlights the use of detachable and autoclavable dispensing cassettes.
Description:
User manual
Environment:
Thermo Scientific Multidrop Combi
Attachment(s):
| File | Last Modified |
|---|---|
| NO5616 - Rev 1.6 - Thermo Scientific Multidrop Combi - User Manual.pdf | December 01, 2021 |