What does the service interval mean?
Issue
What does the service interval mean?
Environment
CO2 Incubator
Resolution
The service interval is a timed reminder to perform the annual maintenance, which includes inspecting the door gasket, replacing the inline CO2 filter, and verifying the calibration of the unit.