7003422 - Series 8000WJ Water Jacketed CO2 Incubator - Operating and Maintenance Manual
The document "7003422 - Series 8000WJ Water Jacketed CO2 Incubator - Operating and Maintenance Manual" provides operational and maintenance guidelines for the Thermo Scientific Series 8000WJ Water Jacketed CO2 Incubator. It includes model-specific information on CO2 sensors (thermal conductivity or infra-red), voltage specifications (115V or 230V), and the presence of O2 sensors.
Description:
Operating and maintenance manual
Environment:
Thermo Scientific Series 8000WJ Water Jacketed CO2 Incubator, models:
| Model | CO2 Sensor* | Voltage** | O2 |
|
IR | 115 | no |
|
IR | 230 | no |
|
T/C | 115 | yes |
|
T/C | 230 | yes |
|
IR | 115 | yes |
|
IR | 230 | yes |
|
T/C | 115 | no |
|
T/C | 230 | no |
*TC is a thermal conductivity sensor. IR is an infra-red sensor.
**All units are 50-60 Hz.