7003110 - Forma Model 3100, 3200 Series - Operating and Maintenance Manual
Description:
Operating and maintenance manual
Environment:
Forma Model 3100, 3200 Series, models:
- 3110 Series Single Chamber Water Jacket Incubator
- 3210 Series Dual Chamber Water Jacket Incubator
Single Chamber Models | |||
Model | CO2 Sensor** | O2 | Voltage*** |
|
T/C | No | 115 |
|
T/C | No | 230 |
|
IR | No | 115 |
|
IR | No | 230 |
|
T/C | Yes | 115 |
|
T/C | Yes | 230 |
|
IR | Yes | 115 |
|
IR | Yes | 230 |
Dual Chamber Models | |||
Model* | CO2 Sensor** | O2 | Voltage*** |
|
T/C | No | 115 |
|
T/C | No | 230 |
|
IR | No | 115 |
|
IR | No | 230 |
|
T/C | Yes | 115 |
|
T/C | Yes | 230 |
|
IR | Yes | 115 |
|
IR | Yes | 230 |
*The models listed as dual chamber models are stacked or dual chamber units. These units arrive as single stand-alone units. They may be stacked one on top of the other or configured side by side with opposite door swing.
**TC is a thermal conductivity sensor. IR is an infrared sensor.
***All units are 50-60 HZ.
Attachment(s):
File | Last Modified |
---|---|
7003110 - Rev 4 - Forma Model 3100_3200 Series - Operating and Maintenance Manual.pdf | December 27, 2021 |
7003110 - Rev 3 - Forma Model 3100_3200 Series - Operating and Maintenance Manual.pdf | December 27, 2021 |
7003110 - Rev 2 - Forma Model 3100_3200 Series - Operating and Maintenance Manual.pdf | December 27, 2021 |
7003110 - Rev 5 - Forma Model 31003200 Series - Operating and Maintenance Manual.pdf | December 27, 2021 |