7004100 - Forma Series 3 Water Jacket CO2 Incubator, Model 4100 Series - Operating Manual REV 15
Description:
Operating manual
Environment:
Forma Series 3 Water Jacket CO2 Incubator, Model 4100 Series:
Single Chamber Models | |||
Model | CO2 Sensor* | O2 | Voltage** |
|
TC | No | 115 |
|
TC | No | 230 |
|
IR | No | 115 |
|
IR | No | 230 |
|
TC | Yes | 115 |
|
TC | Yes | 230 |
|
IR | Yes | 115 |
|
IR | Yes | 230 |
*TC is a thermal conductivity sensor. IR is an infrared sensor.
**All units are 50-60Hz