7010310 - Rev 20 - Forma Direct Heat CO2 Incubator, Model 310 Series - Operating and Maintenance Manual
Description:
Operating and maintenance manual
Environment:
Forma Direct Heat CO2 Incubator, Model 310 Series, models:
Model | CO2 Sensor* | Voltage** | Temp °C Max |
|
T/C | 115 | 55 |
|
T/C | 230 | 55 |
|
IR | 115 | 50 |
|
IR | 230 | 50 |
|
T/C | 115 | 45 |
|
T/C | 230 | 45 |
|
IR | 115 | 45 |
|
IR | 230 | 45 |
* TC is a thermal conductivity sensor. IR is an infrared sensor.
** All units are 50-60 Hz.
† Copper interior
Attachment(s):
File | Last Modified |
---|---|
7010310 - Rev 20 - Forma Direct Heat CO2 Incubator, Model 310 Series - Operating and Maintenance Manual.pdf | December 22, 2021 |