7010310 - Rev 20 - Forma Direct Heat CO2 Incubator_Model 310 Series - Operating and Maintenance Manual
The purpose of the document "7010310 - Rev 20 - Forma Direct Heat CO2 Incubator, Model 310 Series - Operating and Maintenance Manual" is to provide detailed instructions for operating and maintaining the Forma Direct Heat CO2 Incubator, Model 310 Series. It covers various models within this series and includes guidelines on proper usage, maintenance procedures, and troubleshooting tips to ensure optimal performance and longevity of the equipment.
Description:
Operating and maintenance manual
Environment:
Forma Direct Heat CO2 Incubator, Model 310 Series, models:
| Model | CO2 Sensor* | Voltage** | Temp °C Max |
|
T/C | 115 | 55 |
|
T/C | 230 | 55 |
|
IR | 115 | 50 |
|
IR | 230 | 50 |
|
T/C | 115 | 45 |
|
T/C | 230 | 45 |
|
IR | 115 | 45 |
|
IR | 230 | 45 |
* TC is a thermal conductivity sensor. IR is an infrared sensor.
** All units are 50-60 Hz.
† Copper interior
Attachment(s):
| File | Last Modified |
|---|---|
| 7010310 - Rev 20 - Forma Direct Heat CO2 Incubator, Model 310 Series - Operating and Maintenance Manual.pdf | December 22, 2021 |