7010370 - Steri-Cycle CO2 Incubator_Model 370 Series_Direct Heat with Sterilization Cycle - Operating and Maintenance Manual
The document titled "Steri-Cycle CO2 Incubator_Model 370 Series_Direct Heat with Sterilization Cycle - Operating and Maintenance Manual" provides comprehensive guidelines for the operation and maintenance of Steri-Cycle CO2 Incubators, specifically for model numbers 370, 371, 380, and 381. This manual helps users ensure proper functioning and upkeep of these incubators, including instructions on the sterilization cycle feature.
Description:
Operating and maintenance manual
Environment:
Steri-Cycle CO2 Incubator, Model 370 Series, Direct Heat with Sterilization Cycle, model numbers:
| Model | Sensor* | Voltage** |
|
TC | 115 |
|
TC | 230 |
|
IR | 115 |
|
IR | 230 |
*TC is a thermal conductivity sensor. IR is an infra-red sensor.
**All units are 50-60 Hz.