751014991 - Rev 0 - VWR Air Jacketed CO2 Incubator - Operating Instructions
The document titled "751014991 - Rev 0 - VWR Air Jacketed CO2 Incubator - Operating Instructions" provides operating instructions for VWR Air Jacketed CO2 Incubators. It includes specifications such as sensor type, capacity (6.5 cu ft and 6.5 x 2 cu ft), and voltage (115V/60Hz) for the following models: VWR51014991 (TC, 6.5 cu ft) VWR51014992 (IR, 6.5 cu ft) VWR50114902 (TC, 6.5 x 2 cu ft) VWR50114903 (IR, 6.5 x 2 cu ft) This manual is essential for understanding the operation and maintenance o
Description:
Operating instructions
Environment:
VWR Air Jacketed CO2 Incubator, models:
| VWR Catalogue Number | Manufacturer Model Number | Sensor* | Capacity (cu ft) | Voltage |
| 10810-902 |
|
TC | 6.5 | 115V/60Hz |
| 10810-944 |
|
IR | 6.5 | 115V/60Hz |
| 10811-002 |
|
TC | 6.5 X 2** | 115V/60Hz |
| 10811-004 |
|
IR | 6.5 x 2** | 115V/60Hz |
*TC is a thermal conductivity sensor. IR is an infra-red sensor.
**Dual stack
Attachment(s):
| File | Last Modified |
|---|---|
| 751014991 - Rev 0 - VWR Air Jacketed CO2 Incubator - Operating Instructions.pdf | December 22, 2021 |