751014995 - Rev 0 - VWR Water Jacketed CO2 Incubator - Operating Instructions
The document titled "751014995 - Rev 0 - VWR Water Jacketed CO2 Incubator - Operating Instructions" provides operating instructions for various models of VWR Water Jacketed CO2 Incubators. It includes specifications such as sensor type, capacity (6.5 cu ft and 6.5 x 2 cu ft), and voltage (115V/60Hz) for the following models: VWR Catalogue Number 10810-744, Model Number VWR51014995, Sensor TC VWR Catalogue Number 10810-878, Model Number VWR51014996, Sensor IR VWR Catalogue Number 10810-884, Mode
Description:
Operating instructions
Environment:
VWR Water Jacketed CO2 Incubator:
| VWR Catalogue Number | Manufacturer Model Number | Sensor* | Capacity(cu ft) | Voltage |
| 10810-744 |
|
TC | 6.5 | 115V/60Hz |
| 10810-878 |
|
IR | 6.5 | 115V/60Hz |
| 10810-884 |
|
TC | 6.5 x 2** | 115V/60Hz |
| 10810-886 |
|
IR | 6.5 x 2** | 115V/60Hz |
*TC is a thermal conductivity sensor. IR is an infra-red sensor.
**Dual stack
All units include HEPA filtration
Attachment(s):
| File | Last Modified |
|---|---|
| 751014995 - Rev 0 - VWR Water Jacketed CO2 Incubator - Operating Instructions.pdf | December 22, 2021 |