Thermo Fisher Scientific
Error E-29
Error E-29 indicates that upon centrifuge startup, the rotor did not spin or was not identified. To resolve this issue: 1. Ensure there are no obstructions in the chamber and clean the bowl and rotor. 2. Measure motor resistance (2.2 ohms between all phases) and check if the motor shaft spins freely. Replace motor if needed. 3. Verify the rotor ID sensor and ribbon cable integrity. 4. If the error persists, check the Main PCB. 5. Restart the centrifuge.
Issue
Error E-29 signifying that, upon centrifuge start up, the rotor did not spin or was not identified
Environment
Sorvall Legend XTR
Sorvall Legend XT
Sorvall Legend XFR
Sorvall Legend XF
Thermo Scientific SL40R
Thermo Scientific SL40
Sorvall ST40R
Sorvall ST40
Sorvall ST-40R
Sorvall ST-16R
Sorvall ST16
Sorvall ST16R
Sorvall Legend X1R
Sorvall Legend X1
Thermo Scientific SL16R
Thermo Scientific SL16
Thermo Scientific Megafuge 16R
Thermo Scientific Megafuge 16
Thermo Scientific Multifuge X1R
Thermo Scientific Multifuge X1
Thermo Scientific Multifuge X3
Thermo Scientific Multifuge X3R
Thermo Scientific Multifuge X3FR
Thermo Scientific Multifuge X3F
Resolution
Check to make sure that there are no obstructions in the chamber preventing the rotor from spinning free. Also thoroughly clean the entire bowl and rotor. Once verified and clean, measure the resistance of the motor windings for 2.2 ohms between all 3 phases and verify that motor shaft spins free by hand as well. Should the motor need to be replaced, it is P/N 20210429. Keep in mind that a motor grounding kit needs to be installed if it wasn’t previously installed. Contact Technical Support for any assistance or part numbers needed.
Another reason would be rotor identification by the sensor. If we know that the rotor spins upon starting the unit, verify integrity and termination of the ribbon cable from the rotor ID sensor to the main board. The rotor ID sensor is P/N 20150270. Special tool kit P/N 70904693 is needed to remove the auto-lock spindle on the shaft of the motor to access the sensor. If the error persists, the least likely possibility the issue may be with the Main PCB. If no bio-med technician is available to troublehsoot, Field Service Engineer dispatch is recommended.