Overpressure in the Refrigeration Unit
Issue
Overpressure in the refrigeration unit
Environment
Multifuge X Pro-Megafuge ST Plus Centrifuge Series:
| Laboratory Use Centrifuges | In Vitro Diagnostic Centrifuges | ||
| Catalog No. | Benchtop Centrifuge | Catalog No. | Benchtop Centrifuge |
| 75009900 | Multifuge X4 Pro 208-240 V ±10%, 50 / 60 Hz | 75009500 | Multifuge X4 Pro-MD 208-240 V ±10%, 50 / 60 Hz |
| 75009901 | Multifuge X4 Pro 120 V ±10%, 60 Hz | 75009501 | Multifuge X4 Pro-MD 120 V ±10%, 60 Hz |
| 75009915 | Multifuge X4R Pro 220-240 V ±10%, 50 Hz / 230 V ±10%, 60 Hz | 75009515 | Multifuge X4R Pro-MD 220-240 V ±10%, 50 Hz / 230 V ±10%, 60 Hz |
| 75009815 | Multifuge X4R Pro 220 V ±10%, 60 Hz | 75009615 | Multifuge X4R Pro-MD 220 V ±10%, 60 Hz |
| 75009916 | Multifuge X4R Pro 120 V ±10%, 60 Hz | 75009516 | Multifuge X4R Pro-MD 120 V ±10%, 60 Hz |
| 75009903 | Megafuge ST4 Plus 208-240 V ±10%, 50 / 60 Hz | 75009503 | Megafuge ST4 Plus-MD 208-240 V ±10%, 50 / 60 Hz |
| 75009904 | Megafuge ST4 Plus 120 V ±10%, 60 Hz | 75009504 | Megafuge ST4 Plus-MD 120 V ±10%, 60 Hz |
| 75009918 | Megafuge ST4R Plus 220-240 V ±10%, 50 Hz / 230 V ±10%, 60 Hz | 75009518 | Megafuge ST4R Plus-MD 220-240 V ±10%, 50 Hz / 230 V ±10%, 60 Hz |
Resolution
Troubleshooting Tips:
- Clean the air inlet for the condenser.
- Restart the centrifuge.
- If the error message still shows, contact a service technician.