11290085 - Rev A - IEC CL10 - User Manual
The IEC CL10 centrifuge was designed for laboratory use. Through the application of the Relative Centrifugal Force (RCF), it allows the separation of substances made of different density elements. Swing out rotating equipment can generally carry a higher load than a fixed angle or aerodynamical equipment. The larger the diameter of a rotor, the greater the load and the lower the maximum rpm Relative Centrifugal Force (RCF) generated on a centrifuged sample is directly proportional to the sedimen
Description:
Use this manual to get acquainted with the centrifuge and its accessories.
Environment:
IEC CL10
Attachment(s):
| File | Last Modified |
|---|---|
| 11290085 - Rev a - IEC CL10 User Manual.pdf | December 07, 2021 |