Cabinet Leveling
This article provides step-by-step instructions for leveling REVCO series freezers (chest or upright). It emphasizes the importance of a level unit for proper operation and preventing component misalignments. Steps include positioning the freezer in a level, vibration-free area, using a carpenter's level to determine offsets, and adjusting casters or leveling legs accordingly to ensure the unit is level from all sides. Proper leveling ensures optimal performance and longevity of the freezer unit
Issue
Cabinet leveling of a chest or upright freezer
Environment
Revco Series (-3, -5, -7, -9)
Resolution
Refer to the attached document for instructions.
Attachment(s)
| File | Last Modified |
|---|---|
| Cabinet Leveling.pdf | January 13, 2022 |