332192H01 - Rev A -20C Flammable Materials Storage Laboratory Freezers TSX Series Installation and Operation Manual
Description:
This manual provides installation and operation instructions for Flammable Material Storage (FMS) laboratory freezers with a preset temperature setpoint of -20°C.
Environment:
-20° C Flammable Materials Storage Laboratory Freezers TSX Series, model TSX2320HA
Unit Specifications:
| Model | Rated Voltage | Rated Current | Frequency / Phase | Power Module Plug | Approximate Lab Freezer Weight Kg (lbs) | Approximate Freezer Weight Kg (lbs) | Exterior Dimensions (D x W x H) | Hydrocarbon |
| 23A | 115 V | 5.56 A | 60 Hz | IEC C19 | 178 (394) | 203 (449) | 93.6 x 71.1 x 199.4 cm (36.9 x 28.0 x 78.5 in) | R290, 145 g |
Attachment(s):
| File | Last Modified |
|---|---|
| 332192H01 - Rev A - -20C Flammable Materials Storage Laboratory Freezers TSX Series Installation and Operation Manual.pdf | December 03, 2021 |