34830H53 - Rev B - Thermo Scientific Forma Blood Bank Refrigerators - Installation and Operation Manual
Description:
Installation and operation manual
Environment:
Thermo Scientific Forma Blood Bank Refrigerators. This manual provides installation and operation instructions for undercounter models, space saver models and upright models:
| Voltage Codea | Amps/Breaker | Cu. Ft./Liters | Cabinet |
| A | 6.0/15 | 4.7/133 | Undercounter |
| D | 6.0/15 | 4.7/133 | Undercounter |
| V, W | 6.0/15 | 4.7/133 | Undercounter |
| A | 9.4/15 | 11.5/326 | Space Saver |
| D | 6.0/15 | 11.5/326 | Space Saver |
| V, W | 6.0/15 | 11.5/326 | Space Saver |
| A | 9.4/15 | 23.3/659 | Upright |
| D | 6.0/15 | 23.3/659 | Upright |
| V, W | 6.0/15 | 23.3/659 | Upright |
| A | 9.4/15 | 29.2/826 | Upright |
| D | 6.0/15 | 29.2/826 | Upright |
| V, W | 6.0/15 | 29.2/826 | Upright |
| A | 15.0/20 | 51.1/1447 | Upright |
| D | 8.5/15 | 51.1/1447 | Upright |
| V, W | 8.5/15 | 51.1/1447 | Upright |
a V = 230V/50Hz CEE7/7 Plug Cap (Schuko)
W = 230V/50Hz BS1363a Plug Cap
| FRBB404 | FRBB1204 | FRBB2304 | FRBB3004 | FRBB5004 |
| FRBB404A | FRBB1204A | FRBB2304A | FRBB3004A | FRBB5004A |
| FRBB404D | FRBB1204D | FRBB2304D | FRBB3004D |
Attachment(s):
| File | Last Modified |
|---|---|
| 34830H53 - Rev B - Thermo Scientific Forma Blood Bank Refrigerators - Installation and Operation Manual.pdf | December 10, 2021 |