34830H58 - Thermo Scientific Forma Blood Bank Refrigerators - Installation and Operation Manual
Description:
Installation and operation manual
Environment:
Thermo Scientific Forma Blood Bank Refrigerators. This manual provides installation and operation instructions for undercounter models, space saver models and upright models:
| Voltage Codea | Amps/ Breaker | Cu. Ft./ Liters |
Cabinet |
| A | 6.0/15 | 4.7/ | Undercounter |
| D | 6.0/15 | 4.7/ | Undercounter |
| V, W | 6.0/15 | 4.7/ | Undercounter |
| A | 9.4/15 | 11.5/326 | Space Saver |
| D | 6.0/15 | 11.5/326 | Space Saver |
| V, W | 6.0/15 | 11.5/326 | Space Saver |
| A | 9.4/15 | 23.3/659 | Upright |
| D | 6.0/15 | 23.3/659 | Upright |
| V, W | 6.0/15 | 23.3/659 | Upright |
| A | 9.4/15 | 29.2/826 | Upright |
| D | 6.0/15 | 29.2/826 | Upright |
| V, W | 6.0/15 | 29.2/826 | Upright |
| A | 15.0/20 | 51.1/1447 | Upright |
| D | 8.5/15 | 51.1/1447 | Upright |
| V, W | 8.5/15 | 51.1/1447 | Upright |
a A = 115V, 60Hz; D = 208/230V, 60Hz; V = 220V, 50Hz; W = 240V, 50 Hz
| FRBB1204 | FRBB404 |
| FRBB1204D | FRBB404V |
| FRBB1204V | FRBB404W |
| FRBB1204W | FRBB5004 |
| FRBB2304 | FRBB5004D |
| FRBB2304D | FRBB5004V |
| FRBB2304V | FRBB5004W |
| FRBB2304W | FRBB3004V |
| FRBB3004 | FRBB3004W |
| FRBB3004D |
Attachment(s):
| File | Last Modified |
|---|---|
| 34830H58 - Thermo Scientific Forma Blood Bank Refrigerators - Installation and Operation Manual.pdf | December 10, 2021 |