326629H02 - Rev B - Jewett Pass Thru Laboratory Refrigerators - Installation and Operation Manual
Description:
Installation and operation manual
Environment:
Thermo Scientific Jewett Pass-Thru Laboratory Refrigerators, models:
| Model | Voltage | Frequency | Current |
| LR25D-1B | 120V | 60HZ | 9A |
| LR25D-2A* | 230V | 50HZ | 4A |
| LR55D-1B | 120V | 60HZ | 12A |
| LR55D-2A* | 230V | 50HZ | 6.1A |
* Not CE marked
Attachment(s):
| File | Last Modified |
|---|---|
| 326629H02 - Rev B - Jewett Pass Thru Laboratory Refrigerators - Installation and Operation Manual.pdf | December 02, 2021 |