34830H54 - Thermo Scientific Jewett Blood Bank Refrigerators - Installation and Operation Manual
Description:
Installation and operation manual
Environment:
Thermo Scientific Jewett Blood Bank Refrigerators. This manual provides installation and operation instructions for undercounter models, space saver models and upright models:
| Model | Voltage Codea | Amps/ Breaker | Cu. Ft./Liters | Cabinet |
| JBB404 | A | 6.0/15 | 4.7/133 | Undercounter |
| JBB404 | D | 6.0/15 | 4.7/133 | Undercounter |
| JBB404 | V, W | 6.0/15 | 4.7/133 | Undercounter |
| JBB1204 | A | 9.4/15 | 11.5/326 | Space Saver |
| JBB1204 | D | 6.0/15 | 11.5/326 | Space Saver |
| JBB1204 | V, W | 6.0/15 | 11.5/326 | Space Saver |
| JBB2304 | A | 9.4/15 | 23.3/659 | Upright |
| JBB2304 | D | 6.0/15 | 23.3/659 | Upright |
| JBB2304 | V, W | 6.0/15 | 23.3/659 | Upright |
| JBB3004 | A | 9.4/15 | 29.2/826 | Upright |
| JBB3004 | D | 6.0/15 | 29.2/826 | Upright |
| JBB3004 | V, W | 6.0/15 | 29.2/826 | Upright |
| JBB5004 | A | 15.0/20 | 51.1/1447 | Upright |
| JBB5004 | D | 8.5/15 | 51.1/1447 | Upright |
| JBB5004 | V, W | 8.5/15 | 51.1/1447 | Upright |
a V = 230V/50Hz CEE7/7 Plug Cap (Schuko)
W = 230V/50Hz BS1363a Plug Cap