34830H59 - Thermo Scientific Jewett Blood Bank Refrigerators V and W Models - Installation and Operation
Description:
Installation and operation manual
Environment:
Thermo Scientific Jewett Blood Bank Refrigerators, V and W models:
| Model | Voltage Code* | Amps / Breaker | Cu.Ft./ Liters | Cabinet | Door |
| JBB404 | V, W | 6.0/15 | 4.9/133 | Undercounter | Single solid |
| JBB1204 | V, W | 6.0/15 | 11.5/326 | Space Saver | Single hinged glass |
| JBB2304 | V, W | 6.0/15 | 23.3/659 | Upright | Single hinged glass |
| JBB3004 | V, W | 6.0/15 | 29.2/826 | Upright | Single hinged glass |
| JBB5004 | V, W | 8.5/15 | 51.1/1447 | Upright | Double hinged glass |
* V = 230V/50Hz CEE7/7 Plug Cap (Schuko)
W = 230V/50Hz BS1363a Plug Cap