Cryogenic Products - LN2 Tank Supply Pressure Recommendation
Description:
When hooking Cryogenics LN2 Equipment for use it is recommended to have incoming supply pressure at 22 PSI. This pressure level has been found to be the best setting for unit to run efficiently and meet specifications.
Environment:
Models:
| CryoMed Freezer | Control Rate Freezer |
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
Attachment(s):
| File | Last Modified |
|---|---|
| Cryogenic Products - LN2 Tank Supply Pressure Recommendation.pdf | December 09, 2021 |