Cryogenic Storage Vessels - Inaccurate Level Monitor Readings
Description:
This document provides a description of how ice or frost buildup on the transducer or fog in the top part (neck area) of cryogenic vessel contributes to inaccurate level monitor readings.
Environment:
Cryogenic Storage Vessels with Ultrasonic Level Monitor:
- Locator
- Locator Plus
| Locator Jr | CY509105 | 8201 |
| Locator Jr China | CY509105CN | 8202 |
| Locator 4 | CY509107 | 8246 |
| Locator 4 China | CY509107CN | 8247 |
| Locator 6 | Cy5019113 | 8198 |
| Locator 8 | CY509110 | 8211 |
| Locator 8 China | CY509110CN | 8212 |
| Locator Jr Plus | CY509106 | 8237 |
| Locator Jr Plus China | Cy509106CN | 8238 |
| Locator 4 Plus | CY509108 | 8207 |
| Locator 4 Plus China | CY509108CN | 8208 |
| Locator 6 Plus | CY509109 | 8209 |
| Locator 6 Plus China | CY509109CN | 8210 |
| Locator 8 Plus | CY509111 | 8213 |
| Locator 8 Plus China | CY509111CN | 8124 |
Attachment(s):
| File | Last Modified |
|---|---|
| Cryogenic Storage Vessels - Inaccurate Level Monitor Readings.pdf | January 20, 2022 |