Thermo Sciettific Cryopreservation Equipment - Cryopreservation LN2 Supply Dual Setup Instruction Sheet
Description:
It is possible to connect two cryogenic auto-fill units by using a T-valve assembly (Part number: 4000290) or a regular T-fitting provided by local LN2 supplier.
Environment:
CryoExtra and CryoPlus Models Affected:
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
Attachment(s):
| File | Last Modified |
|---|---|
| Thermo Sciettific Cryopreservation Equipment - Cryopreservation LN2 Supply Dual Setup Instruction Sheet.pdf | December 09, 2021 |