Thermo Scientific TSX TLE RLE HLE 89000 ULT Freezers - How to Clear Cloud Data Instruction Sheet
This article provides a step-by-step guide on how to reset cloud data for TSX and Advanced series freezers when Wi-Fi networks are not visible or selectable due to corrupt security certificates. The process involves logging into the freezer with a service username and daily-changing password, navigating to the service settings, and holding the Cloud Data button for 5 seconds to reset the data, which will reboot the User Interface and reconnect the freezer to the cloud as a new device.
Description:
How to reset and clear the UI Assy cloud data for Wi Fi internet connection to Instrument Health.
Environment:
TSX Series, TLE, RLE, HLE, 89000 Advanced ULT Freezers
| 8930086A | HLE30086A | RLE30086A | TLE30086A | TSX40086A |
| 8930086D | HLE30086D | RLE30086D | TLE30086D | TSX40086D |
| 8930086V | HLE30086V | RLE30086V | TLE30086V | TSX40086V |
| 8940086A | HLE40086A | RLE40086A | TLE40086A | TSX40086G |
| 8940086D | HLE40086D | RLE40086D | TLE40086D | TSX50086A |
| 8940086V | HLE40086V | RLE40086V | TLE40086V | TSX50086D |
| 8950086A | HLE50086A | RLE50086A | TLE50086A | TSX50086V |
| 8950086D | HLE50086D | RLE50086D | TLE50086D | TSX50086G |
| 8950086V | HLE50086V | RLE50086V | TLE50086V | TSX60086A |
| 8960086A | HLE60086A | RLE60086A | TLE60086A | TSX60086D |
| 8960086D | HLE60086D | RLE60086D | TLE60086D | TSX60086V |
| 89600V | HLE60086V | RLE60086V | TLE60086V | TSX60086G |
| TSX70086A | ||||
| TSX70086D | ||||
| TSX70086G | ||||
| TSX70086V |
Attachment(s):
| File | Last Modified |
|---|---|
| Thermo Scientific TSX TLE RLE HLE 89000 ULT Freezers - How to Clear Cloud Data Instruction Sheet.pdf | November 22, 2021 |